Learn More
Meso Scale Discovery UPLEX HUM IL1 ASY384well
SDP
Catalog No. 5000080177
Encompass
The Meso Scale Discovery U-PLEX Human IL-1alpha 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of the human cytokine interleukin 1 alpha (IL-1alpha) It utilizes MSD s proprietary electrochemiluminescent detection technology and is designed for use on the MESO QuickPlex SQ 120 instrument The assay has a detection range of 0 05-500 pg/mL and includes a 384-well plate pre-coated with capture antibodies detection antibodies and calibrators It provides high sensitivity requires low sample volume (25 L) and has a short streamlined workflow The kit contains sufficient reagents for one 384-well plate
Catalog No. 50-000-80177 Supplier Meso Scale Discovery Supplier No. K251UNK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-1alpha 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of the human cytokine interleukin 1 alpha (IL-1alpha) It utilizes MSD s proprietary electrochemiluminescent detection technology and is designed for use on the MESO QuickPlex SQ 120 instrument The assay has a detection range of 0 05-500 pg/mL and includes a 384-well plate pre-coated with capture antibodies detection antibodies and calibrators It provides high sensitivity requires low sample volume (25 L) and has a short streamlined workflow The kit contains sufficient reagents for one 384-well plate