Learn More
Meso Scale Discovery UPLEX HUM IL12/IL23p40 ASY384
SDP
Catalog No. 5000081126
Encompass
The Meso Scale Discovery U-PLEX Human IL-12/IL-23p40 384-Well Assay Kit is a multiplex immunoassay that measures IL-12p40 and IL-23 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls It allows simultaneous quantification of IL-12 and IL-23 from a single small sample volume The kit has high performance and is designed for use on the MSD QUICKPLEX SQ 120
Catalog No. 50-000-81126 Supplier Meso Scale Discovery Supplier No. K251UQK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-12/IL-23p40 384-Well Assay Kit is a multiplex immunoassay that measures IL-12p40 and IL-23 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls It allows simultaneous quantification of IL-12 and IL-23 from a single small sample volume The kit has high performance and is designed for use on the MSD QUICKPLEX SQ 120