Learn More
Meso Scale Discovery UPLEX HUM IL12/IL23p40 ASYQP
SDP
Catalog No. 5000078245
Encompass
The U-PLEX Human IL-12/IL-23p40 Assay (K151UQK) is a multiplex immunoassay kit from Meso Scale Discovery that simultaneously measures interleukin-12 and the p40 subunit shared by IL-12 and IL-23 in human samples This electrochemiluminescence-based assay provides high sensitivity detection with minimal sample volume requirements featuring a 96-well plate format for efficient processing of multiple samples The kit includes all necessary reagents and is designed for research applications in immunology and inflammation studies
Catalog No. 50-000-78245 Supplier Meso Scale Discovery Supplier No. K151UQK22
The U-PLEX Human IL-12/IL-23p40 Assay (K151UQK) is a multiplex immunoassay kit from Meso Scale Discovery that simultaneously measures interleukin-12 and the p40 subunit shared by IL-12 and IL-23 in human samples This electrochemiluminescence-based assay provides high sensitivity detection with minimal sample volume requirements featuring a 96-well plate format for efficient processing of multiple samples The kit includes all necessary reagents and is designed for research applications in immunology and inflammation studies