Learn More
Meso Scale Discovery UPLEX HUM IL12/IL23p40 ASYQP
SDP
Catalog No. 5000080397
Encompass
The U-PLEX Human IL-12/IL-23p40 Assay (K151UQK) is a multiplex immunoassay from Meso Scale Discovery for quantitative measurement of interleukin-12 and interleukin-23p40 cytokines in human biological samples This electrochemiluminescence-based assay utilizes proprietary MULTI-SPOT technology for sensitive accurate detection with minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and controls for reliable results in research applications studying inflammatory responses and immune system function
Catalog No. 50-000-80397 Supplier Meso Scale Discovery Supplier No. K151UQK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human IL-12/IL-23p40 Assay (K151UQK) is a multiplex immunoassay from Meso Scale Discovery for quantitative measurement of interleukin-12 and interleukin-23p40 cytokines in human biological samples This electrochemiluminescence-based assay utilizes proprietary MULTI-SPOT technology for sensitive accurate detection with minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and controls for reliable results in research applications studying inflammatory responses and immune system function