Learn More
Meso Scale Discovery UPLEX HUM IL12p70 ASY384well
SDP
Catalog No. 5000079411
Encompass
The Meso Scale Discovery U-PLEX Human IL-12p70 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 12 (IL-12) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantification of IL-12p70 with a lower limit of detection of 0 74 pg/mL and an upper limit of quantification of 2500 pg/mL The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent The total assay time is 3 5 hours
Catalog No. 50-000-79411 Supplier Meso Scale Discovery Supplier No. K251UAK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-12p70 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 12 (IL-12) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantification of IL-12p70 with a lower limit of detection of 0 74 pg/mL and an upper limit of quantification of 2500 pg/mL The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and assay diluent The total assay time is 3 5 hours