Learn More
Meso Scale Discovery UPLEX HUM IL13 ASYQP 25 PL
SDP
Catalog No. 5000079002
Encompass
The U-PLEX Human IL-13 Assay (K151UBK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-13 in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-13 quantification in research applications
Catalog No. 50-000-79002 Supplier Meso Scale Discovery Supplier No. K151UBK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human IL-13 Assay (K151UBK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-13 in human biological samples This electrochemiluminescence-based assay offers high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-13 quantification in research applications