Learn More
Meso Scale Discovery UPLEX HUM IL15 ASYQP 25 PL
SDP
Catalog No. 5000080430
Encompass
The U-PLEX Human IL-15 Assay (K151URK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-15 protein levels in biological samples This electrochemiluminescence-based assay offers high sensitivity detection with minimal sample volume requirements featuring a 96-well plate format for efficient processing The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-15 quantification in research applications
Catalog No. 50-000-80430 Supplier Meso Scale Discovery Supplier No. K151URK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human IL-15 Assay (K151URK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-15 protein levels in biological samples This electrochemiluminescence-based assay offers high sensitivity detection with minimal sample volume requirements featuring a 96-well plate format for efficient processing The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-15 quantification in research applications