Learn More
Meso Scale Discovery UPLEX HUM IL16 ASY384well
SDP
Catalog No. 5000080703
Encompass
The Meso Scale Discovery U-PLEX Human IL-16 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of the human cytokine interleukin 16 (IL-16) in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a 384-well plate format allowing high-throughput analysis of up to 80 samples per plate The sensitivity of the assay ranges from 0 04-1500 pg/mL with an incubation time of 2 hours This quantitative IL-16 immunoassay provides a reproducible and sensitive method for studying immune system function and inflammation
Catalog No. 50-000-80703 Supplier Meso Scale Discovery Supplier No. K251USK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-16 384-Well Assay is a multiplex immunoassay kit that enables simultaneous quantification of the human cytokine interleukin 16 (IL-16) in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a 384-well plate format allowing high-throughput analysis of up to 80 samples per plate The sensitivity of the assay ranges from 0 04-1500 pg/mL with an incubation time of 2 hours This quantitative IL-16 immunoassay provides a reproducible and sensitive method for studying immune system function and inflammation