Learn More
Meso Scale Discovery UPLEX HUM IL17A ASY384well
SDP
Catalog No. 5000079414
Encompass
The Meso Scale Discovery U-PLEX Human IL-17A 384-Well Assay Kit is a multiplex immunoassay that measures IL-17A levels in human serum plasma and cell culture supernatant samples The kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume It includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent The kit can test up to 384 samples in a single run and has a testing time of approximately 3 5 hours
Catalog No. 50-000-79414 Supplier Meso Scale Discovery Supplier No. K251UTK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-17A 384-Well Assay Kit is a multiplex immunoassay that measures IL-17A levels in human serum plasma and cell culture supernatant samples The kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume It includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent The kit can test up to 384 samples in a single run and has a testing time of approximately 3 5 hours