Learn More
Meso Scale Discovery UPLEX HUM IL17A/F ASY 25 PL
SDP
Catalog No. 5000081730
Encompass
The Meso Scale Discovery U-PLEX Human IL-17A/F Assay Kit is a multiplex immunoassay that measures human interleukin 17A and interleukin 17F in cell culture supernatants serum plasma and other biological samples This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 7 and calibrator This assay allows for the simultaneous quantitative measurement of IL-17A and IL-17F from a single sample in as little as 3 5 hours
Catalog No. 50-000-81730 Supplier Meso Scale Discovery Supplier No. K151VYK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-17A/F Assay Kit is a multiplex immunoassay that measures human interleukin 17A and interleukin 17F in cell culture supernatants serum plasma and other biological samples This electrochemiluminescent assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 7 and calibrator This assay allows for the simultaneous quantitative measurement of IL-17A and IL-17F from a single sample in as little as 3 5 hours