Learn More
Meso Scale Discovery UPLEX HUM IL17A/F ASY384well
SDP
Catalog No. 5000081085
Encompass
The Meso Scale Discovery U-PLEX Human IL-17A/F 384-Well Assay is a multiplex immunoassay that measures IL-17A and IL-17F in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer Diluent 2 and Calibrator Blend The assay takes 3 5 hours to complete after a 2 hour incubation
Catalog No. 50-000-81085 Supplier Meso Scale Discovery Supplier No. K251VYK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-17A/F 384-Well Assay is a multiplex immunoassay that measures IL-17A and IL-17F in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer Diluent 2 and Calibrator Blend The assay takes 3 5 hours to complete after a 2 hour incubation