Learn More
Meso Scale Discovery UPLEX HUM IL17A/F ASYQP
SDP
Catalog No. 5000081959
Encompass
The U-PLEX Human IL-17A/F Assay (K151VYK) is a multiplex immunoassay kit from Meso Scale Discovery that simultaneously measures interleukin-17A and IL-17F cytokine levels in human biological samples This electrochemiluminescence-based assay provides high sensitivity detection with minimal sample volume requirements featuring a 96-well plate format for efficient processing of multiple samples The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for quantitative analysis
Catalog No. 50-000-81959 Supplier Meso Scale Discovery Supplier No. K151VYK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human IL-17A/F Assay (K151VYK) is a multiplex immunoassay kit from Meso Scale Discovery that simultaneously measures interleukin-17A and IL-17F cytokine levels in human biological samples This electrochemiluminescence-based assay provides high sensitivity detection with minimal sample volume requirements featuring a 96-well plate format for efficient processing of multiple samples The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for quantitative analysis