Learn More
Meso Scale Discovery UPLEX HUM IL17B ASY 25 PL
SDP
Catalog No. 5000081791
Encompass
The Meso Scale Discovery U-PLEX Human IL-17B Assay Kit is a multiplex immunoassay that measures IL-17B levels in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls to generate a standard curve The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81791 Supplier Meso Scale Discovery Supplier No. K151XNK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-17B Assay Kit is a multiplex immunoassay that measures IL-17B levels in human serum plasma and cell culture supernatant samples The assay uses electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG Read Buffer 10X Diluent and calibrators and controls to generate a standard curve The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL