Learn More
Meso Scale Discovery UPLEX HUM IL17B ASY384well
SDP
Catalog No. 5000080563
Encompass
The Meso Scale Discovery U-PLEX Human IL-17B 384-Well Assay measures IL-17B in human serum plasma and cell culture supernatant using electrochemiluminescence detection It offers high sensitivity a 5-log dynamic range and low sample use The kit includes a 384-well plate with capture antibodies detection antibodies with MSD SULFO-TAG buffers diluent and calibrators It has high specificity with less than 0 3% cross-reactivity Results are analyzed with MSD software enabling efficient IL-17B quantification
Catalog No. 50-000-80563 Supplier Meso Scale Discovery Supplier No. K251XNK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-17B 384-Well Assay measures IL-17B in human serum plasma and cell culture supernatant using electrochemiluminescence detection It offers high sensitivity a 5-log dynamic range and low sample use The kit includes a 384-well plate with capture antibodies detection antibodies with MSD SULFO-TAG buffers diluent and calibrators It has high specificity with less than 0 3% cross-reactivity Results are analyzed with MSD software enabling efficient IL-17B quantification