Learn More
Meso Scale Discovery UPLEX HUM IL17B ASY384well
SDP
Catalog No. 5000080561
Encompass
The Meso Scale Discovery U-PLEX Human IL-17B 384-Well Assay is a multiplex immunoassay that measures IL-17B levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires a small sample volume of 25 L and the 384-well plate allows high-throughput analysis The kit includes a 384-well MULTI-ARRAY Plate Detection Antibody Read Buffer Calibrator and Diluent
Catalog No. 50-000-80561 Supplier Meso Scale Discovery Supplier No. K251XNK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-17B 384-Well Assay is a multiplex immunoassay that measures IL-17B levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires a small sample volume of 25 L and the 384-well plate allows high-throughput analysis The kit includes a 384-well MULTI-ARRAY Plate Detection Antibody Read Buffer Calibrator and Diluent