Learn More
Meso Scale Discovery UPLEX HUM IL17B ASYQP 25 PL
SDP
Catalog No. 5000080459
Encompass
The U-PLEX Human IL-17B Assay (K151XNK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-17B protein levels in human biological samples This electrochemiluminescence-based assay utilizes MSD s proprietary MULTI-SPOT and MULTI-ARRAY technology platforms to deliver sensitive precise detection with minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-17B quantification in research applications studying inflammatory responses and immune system function
Catalog No. 50-000-80459 Supplier Meso Scale Discovery Supplier No. K151XNK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human IL-17B Assay (K151XNK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of interleukin-17B protein levels in human biological samples This electrochemiluminescence-based assay utilizes MSD s proprietary MULTI-SPOT and MULTI-ARRAY technology platforms to deliver sensitive precise detection with minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable IL-17B quantification in research applications studying inflammatory responses and immune system function