Learn More
Meso Scale Discovery UPLEX HUM IL17C ASY384well
SDP
Catalog No. 5000081130
Encompass
The Meso Scale Discovery U-PLEX Human IL-17C 384-Well Assay is an electrochemiluminescent immunoassay for quantifying human interleukin 17C (IL-17C) in various biological matrices It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible IL-17C quantitation with a broad dynamic range Requiring only 25 L of sample it detects as low as 0 04 pg/mL The kit includes a 384-well plate with capture antibody detection antibody conjugated to ruthenium buffers blockers calibrators and controls offering a high-throughput method for IL-17C analysis in research
Catalog No. 50-000-81130 Supplier Meso Scale Discovery Supplier No. K251WJK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-17C 384-Well Assay is an electrochemiluminescent immunoassay for quantifying human interleukin 17C (IL-17C) in various biological matrices It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible IL-17C quantitation with a broad dynamic range Requiring only 25 L of sample it detects as low as 0 04 pg/mL The kit includes a 384-well plate with capture antibody detection antibody conjugated to ruthenium buffers blockers calibrators and controls offering a high-throughput method for IL-17C analysis in research