Learn More
Meso Scale Discovery UPLEX HUM IL17C ASY384well
SDP
Catalog No. 5000080721
Encompass
The Meso Scale Discovery U-PLEX Human IL-17C 384-Well Assay is an electrochemiluminescent immunoassay designed for the quantitative measurement of IL-17C in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in a 384-well format The assay has a detection range of 0 05-1000 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG read buffer wash buffer blocker solutions calibrator and controls This assay allows high-throughput multiplexed cytokine profiling with low sample volume requirements
Catalog No. 50-000-80721 Supplier Meso Scale Discovery Supplier No. K251WJK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-17C 384-Well Assay is an electrochemiluminescent immunoassay designed for the quantitative measurement of IL-17C in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in a 384-well format The assay has a detection range of 0 05-1000 pg/mL and includes a plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG read buffer wash buffer blocker solutions calibrator and controls This assay allows high-throughput multiplexed cytokine profiling with low sample volume requirements