Learn More
Meso Scale Discovery UPLEX HUM IL17D ASY384well
SDP
Catalog No. 5000080654
Encompass
The Meso Scale Discovery U-PLEX Human IL-17D 384-Well Assay is a multiplex immunoassay for measuring IL-17D in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection offering high sensitivity a wide dynamic range and low sample use The kit includes a 384-well plate with capture antibodies detection antibodies with MSD SULFO-TAG Read Buffer Diluent and calibrators It has high specificity and minimal cross-reactivity Results are analyzed with MSD software enabling high throughput quantitative IL-17D measurement
Catalog No. 50-000-80654 Supplier Meso Scale Discovery Supplier No. K251XOK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-17D 384-Well Assay is a multiplex immunoassay for measuring IL-17D in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection offering high sensitivity a wide dynamic range and low sample use The kit includes a 384-well plate with capture antibodies detection antibodies with MSD SULFO-TAG Read Buffer Diluent and calibrators It has high specificity and minimal cross-reactivity Results are analyzed with MSD software enabling high throughput quantitative IL-17D measurement