Learn More
Meso Scale Discovery UPLEX HUM IL17D ASY384well
SDP
Catalog No. 5000080693
Encompass
The Meso Scale Discovery U-PLEX Human IL-17D 384-Well Assay is a multiplex immunoassay for measuring human interleukin 17D (IL-17D) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision This 384-well assay enables high throughput analysis in 3 5 hours requiring only 50 L of sample and detecting as low as 0 06 pg/mL It includes a plate with capture antibodies detection antibodies with electrochemiluminescent labels and calibrator and control samples for reliable IL-17D quantitation
Catalog No. 50-000-80693 Supplier Meso Scale Discovery Supplier No. K251XOK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-17D 384-Well Assay is a multiplex immunoassay for measuring human interleukin 17D (IL-17D) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision This 384-well assay enables high throughput analysis in 3 5 hours requiring only 50 L of sample and detecting as low as 0 06 pg/mL It includes a plate with capture antibodies detection antibodies with electrochemiluminescent labels and calibrator and control samples for reliable IL-17D quantitation