Learn More
Meso Scale Discovery UPLEX HUM IL17D ASYQP 25 PL
SDP
Catalog No. 5000079816
Encompass
The U-PLEX Human IL-17D Assay (K151XOK) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure interleukin-17D levels in human biological samples This electrochemiluminescence-based assay utilizes MSD s proprietary MULTI-SPOT and MULTI-ARRAY technology platforms for sensitive and specific cytokine detection The kit provides researchers with validated reagents and protocols for IL-17D analysis in serum plasma and cell culture supernatants supporting inflammation and immunology research applications
Catalog No. 50-000-79816 Supplier Meso Scale Discovery Supplier No. K151XOK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human IL-17D Assay (K151XOK) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure interleukin-17D levels in human biological samples This electrochemiluminescence-based assay utilizes MSD s proprietary MULTI-SPOT and MULTI-ARRAY technology platforms for sensitive and specific cytokine detection The kit provides researchers with validated reagents and protocols for IL-17D analysis in serum plasma and cell culture supernatants supporting inflammation and immunology research applications