Learn More
Meso Scale Discovery UPLEX HUM IL17E/IL25 ASY
SDP
Catalog No. 5000079595
Encompass
The Meso Scale Discovery U-PLEX Human IL-17E/IL-25 Assay Kit is a multiplex immunoassay that measures human IL-17E and IL-25 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator The kit has a typical sensitivity of 0 04 pg/mL for IL-17E and 0 2 pg/mL for IL-25
Catalog No. 50-000-79595 Supplier Meso Scale Discovery Supplier No. K151VZK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-17E/IL-25 Assay Kit is a multiplex immunoassay that measures human IL-17E and IL-25 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator The kit has a typical sensitivity of 0 04 pg/mL for IL-17E and 0 2 pg/mL for IL-25