Learn More
Meso Scale Discovery UPLEX HUM IL17E/IL25 ASY384
SDP
Catalog No. 5000079473
Encompass
The Meso Scale Discovery U-PLEX Human IL-17E/IL-25 384-Well Assay is a multiplex immunoassay kit for quantifying human interleukin 17E (IL-17E) and interleukin 25 (IL-25) using electrochemiluminescent detection It features Meso Scale Discovery s multi-array plate technology offering high sensitivity a 5-log dynamic range and low sample volume needs This 384-well format supports high-throughput analysis of IL-17E and IL-25 crucial in type 2 immunity and allergic inflammation using 50 L of various sample types per well
Catalog No. 50-000-79473 Supplier Meso Scale Discovery Supplier No. K251VZK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-17E/IL-25 384-Well Assay is a multiplex immunoassay kit for quantifying human interleukin 17E (IL-17E) and interleukin 25 (IL-25) using electrochemiluminescent detection It features Meso Scale Discovery s multi-array plate technology offering high sensitivity a 5-log dynamic range and low sample volume needs This 384-well format supports high-throughput analysis of IL-17E and IL-25 crucial in type 2 immunity and allergic inflammation using 50 L of various sample types per well