Learn More
Meso Scale Discovery UPLEX HUM IL17E/IL25 ASYQP
SDP
Catalog No. 5000079750
Encompass
The MSD U-PLEX Human IL-17E/IL-25 Singleplex Assay is an electrochemiluminescence-based immunoassay kit designed for quantitative measurement of interleukin-17E (also known as IL-25) in human biological samples This singleplex assay utilizes Meso Scale Discovery s proprietary U-PLEX platform technology offering high sensitivity detection with minimal sample volume requirements The kit is specifically validated for human specimens and provides researchers with a reliable tool for cytokine analysis in immunology and inflammation studies
Catalog No. 50-000-79750 Supplier Meso Scale Discovery Supplier No. K151VZK21
The MSD U-PLEX Human IL-17E/IL-25 Singleplex Assay is an electrochemiluminescence-based immunoassay kit designed for quantitative measurement of interleukin-17E (also known as IL-25) in human biological samples This singleplex assay utilizes Meso Scale Discovery s proprietary U-PLEX platform technology offering high sensitivity detection with minimal sample volume requirements The kit is specifically validated for human specimens and provides researchers with a reliable tool for cytokine analysis in immunology and inflammation studies