Learn More
Meso Scale Discovery UPLEX HUM IL17F ASY384well
SDP
Catalog No. 5000080513
Encompass
The Meso Scale Discovery U-PLEX Human IL-17F 384-Well Assay is a multiplex immunoassay for measuring IL-17F in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume This 384-well plate assay quantifies IL-17F efficiently in 3 5 hours crucial for studying inflammatory and autoimmune responses The kit includes a 384-well plate with capture antibodies detection antibodies read buffer wash buffer calibrator and controls
Catalog No. 50-000-80513 Supplier Meso Scale Discovery Supplier No. K251WAK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-17F 384-Well Assay is a multiplex immunoassay for measuring IL-17F in human serum plasma and cell culture supernatant It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume This 384-well plate assay quantifies IL-17F efficiently in 3 5 hours crucial for studying inflammatory and autoimmune responses The kit includes a 384-well plate with capture antibodies detection antibodies read buffer wash buffer calibrator and controls