Learn More
Meso Scale Discovery UPLEX HUM IL17F ASY384well
SDP
Supplier:  Meso Scale Discovery K251WAK4
Encompass
The Meso Scale Discovery U-PLEX Human IL-17F 384-Well Assay is a multiplex immunoassay that measures IL-17F levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a small sample volume requirement This 384-well assay allows high throughput analysis of IL-17F an inflammatory cytokine involved in autoimmune diseases It requires an MSD instrument such as SECTOR Imager 6000 for plate reading
Catalog No. 50-000-80975
Only null left
Add to Cart
Request a Quote