Learn More
Meso Scale Discovery UPLEX HUM IL17F ASYQP 25 PL
SDP
Catalog No. 5000079539
Encompass
The U-PLEX Human IL-17F Assay (K151WAK) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure interleukin-17F levels in human biological samples This electrochemiluminescence-based assay utilizes MSD s proprietary MULTI-SPOT and MULTI-ARRAY technology platforms for enhanced sensitivity and precision The kit is suitable for research applications studying inflammatory responses autoimmune diseases and cytokine profiling in serum plasma and cell culture samples
Catalog No. 50-000-79539 Supplier Meso Scale Discovery Supplier No. K151WAK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human IL-17F Assay (K151WAK) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure interleukin-17F levels in human biological samples This electrochemiluminescence-based assay utilizes MSD s proprietary MULTI-SPOT and MULTI-ARRAY technology platforms for enhanced sensitivity and precision The kit is suitable for research applications studying inflammatory responses autoimmune diseases and cytokine profiling in serum plasma and cell culture samples