Learn More
Meso Scale Discovery UPLEX HUM IL18 ASY384well
SDP
Catalog No. 5000079852
Encompass
The Meso Scale Discovery U-PLEX Human IL-18 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 18 (IL-18) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 61 pg/mL This assay is for research use only and not for use in diagnostic procedures It is available as a kit including a 96-well plate coated with capture antibody detection antibody calibrator and assay diluent
Catalog No. 50-000-79852 Supplier Meso Scale Discovery Supplier No. K251VJK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-18 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 18 (IL-18) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 61 pg/mL This assay is for research use only and not for use in diagnostic procedures It is available as a kit including a 96-well plate coated with capture antibody detection antibody calibrator and assay diluent