Learn More
Meso Scale Discovery UPLEX HUM IL1RA ASY 25 PL
SDP
Catalog No. 5000081812
Encompass
The Meso Scale Discovery U-PLEX Human IL-1ra Assay Kit is a multiplex immunoassay that measures IL-1ra levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81812 Supplier Meso Scale Discovery Supplier No. K151XPK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-1ra Assay Kit is a multiplex immunoassay that measures IL-1ra levels in human serum plasma and cell culture supernatants The assay uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and assay diluent The assay has a typical sensitivity of 0 08 pg/mL and an upper limit of quantification of 10 000 pg/mL