Learn More
Meso Scale Discovery UPLEX HUM IL1RA ASY384well
SDP
Catalog No. 5000081131
Encompass
The Meso Scale Discovery U-PLEX Human IL-1ra 384-Well Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human interleukin-1 receptor antagonist (IL-1ra) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of IL-1ra with a broad dynamic range and rapid protocol This assay is suitable for biomarker research and drug discovery applications involving IL-1ra The kit includes a 384-well plate coated with capture antibody detection antibody read buffer calibration standard and sample diluent
Catalog No. 50-000-81131 Supplier Meso Scale Discovery Supplier No. K251XPK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-1ra 384-Well Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human interleukin-1 receptor antagonist (IL-1ra) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of IL-1ra with a broad dynamic range and rapid protocol This assay is suitable for biomarker research and drug discovery applications involving IL-1ra The kit includes a 384-well plate coated with capture antibody detection antibody read buffer calibration standard and sample diluent