Learn More
Meso Scale Discovery UPLEX HUM IL1RA ASY384well
SDP
Catalog No. 5000079419
Encompass
The Meso Scale Discovery U-PLEX Human IL-1ra 384-Well Assay Kit is a 384-well multiplex immunoassay that measures human interleukin-1 receptor antagonist (IL-1ra) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 384-well plate coated with capture antibody detection antibody read buffer calibrator and controls The assay has a typical sensitivity of 0 08 pg/mL and requires 25 L sample size per well
Catalog No. 50-000-79419 Supplier Meso Scale Discovery Supplier No. K251XPK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-1ra 384-Well Assay Kit is a 384-well multiplex immunoassay that measures human interleukin-1 receptor antagonist (IL-1ra) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This kit includes a 384-well plate coated with capture antibody detection antibody read buffer calibrator and controls The assay has a typical sensitivity of 0 08 pg/mL and requires 25 L sample size per well