Learn More
Meso Scale Discovery UPLEX HUM IL21 ASY384well
SDP
Catalog No. 5000080650
Encompass
The Meso Scale Discovery U-PLEX Human IL-21 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 21 (IL-21) in cell culture supernatants serum plasma (EDTA heparin citrate) saliva and urine The assay has a sensitivity of 0 03 pg/mL and a 7-log dynamic range from 0 03 - 10 000 pg/mL The kit includes a 384-well MULTI-ARRAY plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and assay diluent The assay takes 3 5 hours to complete after a 2 hour incubation
Catalog No. 50-000-80650 Supplier Meso Scale Discovery Supplier No. K251WBK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-21 384-Well Assay Kit is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 21 (IL-21) in cell culture supernatants serum plasma (EDTA heparin citrate) saliva and urine The assay has a sensitivity of 0 03 pg/mL and a 7-log dynamic range from 0 03 - 10 000 pg/mL The kit includes a 384-well MULTI-ARRAY plate coated with capture antibody detection antibody conjugated to MSD SULFO-TAG calibrators and assay diluent The assay takes 3 5 hours to complete after a 2 hour incubation