Learn More
Meso Scale Discovery UPLEX HUM IL22 ASY384well
SDP
Catalog No. 5000080981
Encompass
The Meso Scale Discovery U-PLEX Human IL-22 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous measurement of IL-22 cytokine in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescence detection technology and provides a 384-well plate formatted assay complete with analyte-specific antibodies pre-coated onto spots on the bottom of the wells The kit includes a 10-plate box providing 3 840 wells that are ready to use off the shelf With high sensitivity broad dynamic range and minimal sample volume requirements this assay kit offers a highly efficient way to quantitatively analyze IL-22
Catalog No. 50-000-80981 Supplier Meso Scale Discovery Supplier No. K251WIK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-22 384-Well Assay is a multiplex immunoassay kit that enables the simultaneous measurement of IL-22 cytokine in human serum plasma and cell culture supernatant samples It utilizes electrochemiluminescence detection technology and provides a 384-well plate formatted assay complete with analyte-specific antibodies pre-coated onto spots on the bottom of the wells The kit includes a 10-plate box providing 3 840 wells that are ready to use off the shelf With high sensitivity broad dynamic range and minimal sample volume requirements this assay kit offers a highly efficient way to quantitatively analyze IL-22