Learn More
Meso Scale Discovery UPLEX HUM IL27 ASY384well
SDP
Catalog No. 5000079418
Encompass
The U-PLEX Human IL-27 (384-well Assay) is an immunoassay that enables the simultaneous detection of multiple biomarkers in human serum plasma and cell culture supernatants This 384-well assay kit contains a plate coated with analyte-specific antibodies detection antibodies and standards to quantify IL-27 levels The electrochemiluminescence immunoassay provides high sensitivity a broad dynamic range and requires low sample volumes This assay kit can help researchers investigate the role of IL-27 in regulating immune responses With a streamlined workflow and minimal hands-on time this kit allows high-throughput analysis of IL-27
Catalog No. 50-000-79418 Supplier Meso Scale Discovery Supplier No. K251WCK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human IL-27 (384-well Assay) is an immunoassay that enables the simultaneous detection of multiple biomarkers in human serum plasma and cell culture supernatants This 384-well assay kit contains a plate coated with analyte-specific antibodies detection antibodies and standards to quantify IL-27 levels The electrochemiluminescence immunoassay provides high sensitivity a broad dynamic range and requires low sample volumes This assay kit can help researchers investigate the role of IL-27 in regulating immune responses With a streamlined workflow and minimal hands-on time this kit allows high-throughput analysis of IL-27