Learn More
Meso Scale Discovery UPLEX HUM IL29 ASY
SDP
Catalog No. 5000078734
Encompass
The Meso Scale Discovery U-PLEX Human IL-29 (IFN Lambda1) Assay Kit is a multiplex immunoassay that measures human IL-29 (IFN Lambda1) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls The assay takes approximately 3 5 hours to run and 10 L of sample is required The lower limit of detection is 0 04 pg/mL
Catalog No. 50-000-78734 Supplier Meso Scale Discovery Supplier No. K151WDK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-29 (IFN Lambda1) Assay Kit is a multiplex immunoassay that measures human IL-29 (IFN Lambda1) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and controls The assay takes approximately 3 5 hours to run and 10 L of sample is required The lower limit of detection is 0 04 pg/mL