Learn More
Meso Scale Discovery UPLEX HUM IL29IFN1 ASYQP
SDP
Catalog No. 5000081927
Encompass
The U-PLEX Human IL-29 (IFN-lambda1) Assay (K151WDK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of Interleukin-29 also known as Interferon-lambda1 in biological samples This electrochemiluminescence-based assay provides high sensitivity detection and can be combined with other U-PLEX assays for simultaneous multi-analyte analysis The kit includes pre-coated plates detection antibodies and calibrators for reliable cytokine quantification in research applications studying immune responses and inflammatory pathways
Catalog No. 50-000-81927 Supplier Meso Scale Discovery Supplier No. K151WDK22
The U-PLEX Human IL-29 (IFN-lambda1) Assay (K151WDK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of Interleukin-29 also known as Interferon-lambda1 in biological samples This electrochemiluminescence-based assay provides high sensitivity detection and can be combined with other U-PLEX assays for simultaneous multi-analyte analysis The kit includes pre-coated plates detection antibodies and calibrators for reliable cytokine quantification in research applications studying immune responses and inflammatory pathways