Learn More
Meso Scale Discovery UPLEX HUM IL2R ASY 25 PL
SDP
Catalog No. 5000081789
Encompass
The Meso Scale Discovery U-PLEX Human IL-2Ralpha Assay is a multiplex immunoassay kit that enables simultaneous quantification of IL-2Ralpha in human serum plasma and cell culture supernatant samples The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the accurate and reproducible measurement of IL-2Ralpha which can aid in understanding immune system function and disease progression With minimal hands-on time required this kit serves as a powerful tool for biomarker discovery and validation
Catalog No. 50-000-81789 Supplier Meso Scale Discovery Supplier No. K151XGK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-2Ralpha Assay is a multiplex immunoassay kit that enables simultaneous quantification of IL-2Ralpha in human serum plasma and cell culture supernatant samples The assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay allows for the accurate and reproducible measurement of IL-2Ralpha which can aid in understanding immune system function and disease progression With minimal hands-on time required this kit serves as a powerful tool for biomarker discovery and validation