Learn More
Meso Scale Discovery UPLEX HUM IL2R ASY384well
SDP
Catalog No. 5000080977
Encompass
The Meso Scale Discovery U-PLEX Human IL-2Ralpha 384-Well Assay is a multiplex immunoassay that measures IL-2Ralpha levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This 384-well assay allows for efficient and high-throughput analysis of IL-2Ralpha It requires only 25 L of sample per well and the plate can be read in approximately 2 hours The assay is validated for research use only not for diagnostic procedures It provides quantitative determination of IL-2Ralpha concentrations
Catalog No. 50-000-80977 Supplier Meso Scale Discovery Supplier No. K251XGK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-2Ralpha 384-Well Assay is a multiplex immunoassay that measures IL-2Ralpha levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption This 384-well assay allows for efficient and high-throughput analysis of IL-2Ralpha It requires only 25 L of sample per well and the plate can be read in approximately 2 hours The assay is validated for research use only not for diagnostic procedures It provides quantitative determination of IL-2Ralpha concentrations