Learn More
Meso Scale Discovery UPLEX HUM IL2R ASY384well
SDP
Catalog No. 5000080620
Encompass
The Meso Scale Discovery U-PLEX Human IL-2Ralpha 384-Well Assay is an electrochemiluminescent immunoassay designed for the quantitative measurement of IL-2Ralpha in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in a 384-well format The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It requires a low sample volume of 25 L and the total assay time is approximately 3 5 hours This product can aid in understanding IL-2Ralpha biology and evaluating immunotherapies targeting the IL-2/IL-2R pathway
Catalog No. 50-000-80620 Supplier Meso Scale Discovery Supplier No. K251XGK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-2Ralpha 384-Well Assay is an electrochemiluminescent immunoassay designed for the quantitative measurement of IL-2Ralpha in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in a 384-well format The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL It requires a low sample volume of 25 L and the total assay time is approximately 3 5 hours This product can aid in understanding IL-2Ralpha biology and evaluating immunotherapies targeting the IL-2/IL-2R pathway