Learn More
Meso Scale Discovery UPLEX HUM IL3 ASY384well
SDP
Catalog No. 5000080724
Encompass
The Meso Scale Discovery U-PLEX Human IL-3 384-Well Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 3 (IL-3) in cell culture supernatants serum plasma and other biological matrices This multiplex assay kit allows for the simultaneous measurement of IL-3 along with other biomarkers in a 384-well plate format The assay has a sensitivity of 0 04 pg/mL and a broad dynamic range from 0 06 - 2500 pg/mL The total assay time is 3 5 hours This assay provides a rapid sensitive and reproducible way to measure IL-3 levels across multiple samples
Catalog No. 50-000-80724 Supplier Meso Scale Discovery Supplier No. K251XMK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-3 384-Well Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 3 (IL-3) in cell culture supernatants serum plasma and other biological matrices This multiplex assay kit allows for the simultaneous measurement of IL-3 along with other biomarkers in a 384-well plate format The assay has a sensitivity of 0 04 pg/mL and a broad dynamic range from 0 06 - 2500 pg/mL The total assay time is 3 5 hours This assay provides a rapid sensitive and reproducible way to measure IL-3 levels across multiple samples