Learn More
Meso Scale Discovery UPLEX HUM IL31 ASY384well
SDP
Catalog No. 5000080180
Encompass
The Meso Scale Discovery U-PLEX Human IL-31 384-Well Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 31 (IL-31) in serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to ruthenium label read buffer 10X diluent and calibrator The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-2500 pg/mL
Catalog No. 50-000-80180 Supplier Meso Scale Discovery Supplier No. K251WEK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-31 384-Well Assay is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 31 (IL-31) in serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results in just 3 5 hours The kit includes a 384-well plate coated with capture antibody detection antibody conjugated to ruthenium label read buffer 10X diluent and calibrator The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-2500 pg/mL