Learn More
Meso Scale Discovery UPLEX HUM IL4 ASY384well
SDP
Catalog No. 5000081042
Encompass
The Meso Scale Discovery U-PLEX Human IL-4 384-Well Assay Kit is a multiplex immunoassay that measures human interleukin 4 (IL-4) in cell culture supernatants serum plasma and other biological fluids This kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay is provided in a 384-well plate format and includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control solutions This allows for the accurate and reliable quantification of IL-4 levels from human samples
Catalog No. 50-000-81042 Supplier Meso Scale Discovery Supplier No. K251TWK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-4 384-Well Assay Kit is a multiplex immunoassay that measures human interleukin 4 (IL-4) in cell culture supernatants serum plasma and other biological fluids This kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay is provided in a 384-well plate format and includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator and control solutions This allows for the accurate and reliable quantification of IL-4 levels from human samples