Learn More
Meso Scale Discovery UPLEX HUM IL5 ASY384well
SDP
Catalog No. 5000080170
Encompass
The Meso Scale Discovery U-PLEX Human IL-5 384-Well Assay is a multiplex immunoassay that measures IL-5 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be run on Meso Scale Discovery s instrumentation platforms and has a testing time of approximately 3 5 hours It includes a 384-well plate pre-coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator The assay measures IL-5 levels between 0 06 - 2500 pg/mL
Catalog No. 50-000-80170 Supplier Meso Scale Discovery Supplier No. K251UOK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-5 384-Well Assay is a multiplex immunoassay that measures IL-5 levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay can be run on Meso Scale Discovery s instrumentation platforms and has a testing time of approximately 3 5 hours It includes a 384-well plate pre-coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator The assay measures IL-5 levels between 0 06 - 2500 pg/mL