Learn More
Meso Scale Discovery UPLEX HUM IL5 ASYQP 25 PL
SDP
Catalog No. 5000079723
Encompass
The U-PLEX Human IL-5 Assay (K151UOK) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure interleukin-5 levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for IL-5 detection supporting research in inflammation allergy and immune response studies The kit provides reliable reproducible results with minimal sample volume requirements
Catalog No. 50-000-79723 Supplier Meso Scale Discovery Supplier No. K151UOK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human IL-5 Assay (K151UOK) is a multiplex immunoassay kit from Meso Scale Discovery designed to quantitatively measure interleukin-5 levels in human biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for IL-5 detection supporting research in inflammation allergy and immune response studies The kit provides reliable reproducible results with minimal sample volume requirements