Learn More
Meso Scale Discovery UPLEX HUM IL7 ASY384well
SDP
Catalog No. 5000079413
Encompass
The Meso Scale Discovery U-PLEX Human IL-7 Assay Kit is a multiplex immunoassay that measures human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This 4-plex assay kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and an MSD read buffer for analysis on MSD instruments The assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-79413 Supplier Meso Scale Discovery Supplier No. K251UPK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-7 Assay Kit is a multiplex immunoassay that measures human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This 4-plex assay kit includes a plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and an MSD read buffer for analysis on MSD instruments The assay has a sensitivity of 0 3 pg/mL and an upper limit of quantification of 10 000 pg/mL