Learn More
Meso Scale Discovery UPLEX HUM IL9 ASY384well
SDP
Catalog No. 5000080661
Encompass
The Meso Scale Discovery U-PLEX Human IL-9 384-Well Assay is a multiplex immunoassay kit for the quantitative measurement of human interleukin 9 (IL-9) in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a 384-well plate format allowing high-throughput analysis of up to 80 samples per plate The assay has a sensitivity of 0 04 pg/mL and a broad dynamic range of up to 10 000 pg/mL The total assay time is approximately 3 5 hours This assay provides a sensitive reproducible method for quantifying IL-9 levels in biological samples
Catalog No. 50-000-80661 Supplier Meso Scale Discovery Supplier No. K251XKK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IL-9 384-Well Assay is a multiplex immunoassay kit for the quantitative measurement of human interleukin 9 (IL-9) in serum plasma and cell culture supernatants The assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a 384-well plate format allowing high-throughput analysis of up to 80 samples per plate The assay has a sensitivity of 0 04 pg/mL and a broad dynamic range of up to 10 000 pg/mL The total assay time is approximately 3 5 hours This assay provides a sensitive reproducible method for quantifying IL-9 levels in biological samples