Learn More
Meso Scale Discovery UPLEX HUM Insulin ASY384well
SDP
Catalog No. 5000081103
Encompass
The Meso Scale Discovery U-PLEX Human Insulin 384-Well Assay Kit is a multiplex immunoassay that measures insulin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 384-well plate assay allows for high-throughput analysis of insulin with minimal sample volume requirements The kit includes a 384-well MULTI-ARRAY plate coated with capture antibodies detection antibodies read buffer calibrators and controls needed to quantitatively determine insulin concentrations
Catalog No. 50-000-81103 Supplier Meso Scale Discovery Supplier No. K2516HK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Insulin 384-Well Assay Kit is a multiplex immunoassay that measures insulin levels in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This 384-well plate assay allows for high-throughput analysis of insulin with minimal sample volume requirements The kit includes a 384-well MULTI-ARRAY plate coated with capture antibodies detection antibodies read buffer calibrators and controls needed to quantitatively determine insulin concentrations