Learn More
Meso Scale Discovery UPLEX HUM Insulin ASYQP
SDP
Catalog No. 5000081694
Encompass
MSD s U-PLEX Human Insulin Singleplex Assay is an electrochemiluminescence-based immunoassay for quantitative measurement of insulin in human biological samples This high-sensitivity assay utilizes MSD s proprietary MULTI-SPOT and MULTI-ARRAY technology platforms offering superior precision and dynamic range compared to traditional ELISA methods The singleplex format allows focused analysis of insulin levels in serum plasma or other sample types making it ideal for diabetes research metabolic studies and clinical diagnostics requiring accurate insulin quantification
Catalog No. 50-000-81694 Supplier Meso Scale Discovery Supplier No. K1516HK24
Add to Cart
Add to Cart
Request a Quote
MSD s U-PLEX Human Insulin Singleplex Assay is an electrochemiluminescence-based immunoassay for quantitative measurement of insulin in human biological samples This high-sensitivity assay utilizes MSD s proprietary MULTI-SPOT and MULTI-ARRAY technology platforms offering superior precision and dynamic range compared to traditional ELISA methods The singleplex format allows focused analysis of insulin levels in serum plasma or other sample types making it ideal for diabetes research metabolic studies and clinical diagnostics requiring accurate insulin quantification