Learn More
Meso Scale Discovery UPLEX HUM IP10 ASY384well
SDP
Catalog No. 5000080615
Encompass
The Meso Scale Discovery U-PLEX Human IP-10 384-Well Assay Kit is a multiplex immunoassay for the quantitative measurement of human interferon gamma-induced protein 10 (IP-10) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology in a 384-well plate format providing high sensitivity and a broad dynamic range The assay requires a low sample volume of 25 L and has a testing time of approximately 3 5 hours The kit includes a 384-well MULTI-ARRAY Plate Detection Antibody Read Buffer Calibrator and Diluent It has a sensitivity of 0 06 pg/mL and a dynamic range of 0 06 - 2500 pg/mL
Catalog No. 50-000-80615 Supplier Meso Scale Discovery Supplier No. K251UFK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human IP-10 384-Well Assay Kit is a multiplex immunoassay for the quantitative measurement of human interferon gamma-induced protein 10 (IP-10) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology in a 384-well plate format providing high sensitivity and a broad dynamic range The assay requires a low sample volume of 25 L and has a testing time of approximately 3 5 hours The kit includes a 384-well MULTI-ARRAY Plate Detection Antibody Read Buffer Calibrator and Diluent It has a sensitivity of 0 06 pg/mL and a dynamic range of 0 06 - 2500 pg/mL