Learn More
Meso Scale Discovery UPLEX HUM ITAC ASY384well
SDP
Catalog No. 5000080613
Encompass
The U-PLEX Human I-TAC 384-Well Assay from Meso Scale Discovery is a 384-well electrochemiluminescent immunoassay designed for the quantitative measurement of human Interferon-inducible T-cell Alpha Chemoattractant (I-TAC) in serum plasma and cell culture supernatants This 10-plex assay has a detection range of 0 06-2500 pg/mL and requires as little as 25 L of sample The assay takes approximately 3 5 hours to complete and utilizes MSD s patented MULTI-ARRAY technology with integrated screen-printed carbon ink electrodes for enhanced sensitivity and reproducibility This assay allows for high-throughput multiplex biomarker quantification with minimal sample consumption
Catalog No. 50-000-80613 Supplier Meso Scale Discovery Supplier No. K251UWK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human I-TAC 384-Well Assay from Meso Scale Discovery is a 384-well electrochemiluminescent immunoassay designed for the quantitative measurement of human Interferon-inducible T-cell Alpha Chemoattractant (I-TAC) in serum plasma and cell culture supernatants This 10-plex assay has a detection range of 0 06-2500 pg/mL and requires as little as 25 L of sample The assay takes approximately 3 5 hours to complete and utilizes MSD s patented MULTI-ARRAY technology with integrated screen-printed carbon ink electrodes for enhanced sensitivity and reproducibility This assay allows for high-throughput multiplex biomarker quantification with minimal sample consumption